Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212513.2 | NP_997678.2 | 4 | 843 | ||
XM_006237348.2 | XP_006237410.1 | 4 | 1068 | |||
XM_008763282.1 | XP_008761504.1 | 5 | 949 | |||
XM_008763283.1 | XP_008761505.1 | 3 | 593 | |||
GenBank mRNA | AY230414.1 | - | 4 | 733 | ||
BC091201.1 | - | 4 | 843 | 7380279 | ||
DQ438937.1 | - | 4 | 721 |