Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001796.4 | NP_001001796.1 | 7 | 1316 | ||
GenBank mRNA | AK039633.1 | - | 7 | 1317 | ||
EU670678.1 | - | 8 | 1436 |