Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd34a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024851.3 | NP_001020022.2 | 2 | 2338 | ||
GenBank mRNA | BC052924.1 | - | 2 | 2318 |