Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033669.2 | NP_001028841.2 | 6, 7 | 1414 | ||
NM_001291441.1 | NP_001278370.1 | 6, 7 | 1414 | |||
GenBank mRNA | AK132680.1 | - | 6, 7 | 1415 | ||
BC141212.1 | - | 6, 7 | 1405 | 9055824 |