Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vrtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033776.2 | NP_001028948.1 | 2 | 2579 | ||
NM_001168588.1 | NP_001162060.1 | 3 | 2991 | |||
NM_001168589.2 | NP_001162061.1 | 2 | 2649 | |||
GenBank mRNA | AK135789.1 | - | 1 | 1181 | ||
BC103771.1 | - | 2 | 2579 | 30935479 |