Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037987.3 | NP_001033076.1 | 9 | 1634 | ||
NM_010103.4 | NP_034233.1 | 8 | 1604 | |||
GenBank mRNA | AF031524.1 | - | 9 | 1742 | ||
AK036117.1 | - | 9 | 1595 | |||
AK046885.1 | - | 9 | 1635 | |||
AK081003.1 | - | 8 | 1436 | |||
AK139093.1 | - | 9 | 1596 | |||
AK141907.1 | - | 9 | 1618 | |||
BC056386.1 | - | 9 | 1534 | 6854497 |