Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-T22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010397.4 | NP_034527.1 | 2 | 527 | ||
GenBank mRNA | AB359229.1 | - | 2 | 272 | ||
AK133985.1 | - | 3 | 459 | |||
AK161744.1 | - | 2 | 526 | |||
AK162874.1 | - | 3 | 429 | |||
BC003819.1 | - | 3 | 406 | 3484613 | ||
M35243.1 | - | 2 | 496 | |||
M35245.1 | - | 2 | 527 |