Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010455.2 | NP_034585.1 | 1 | 1200 | ||
GenBank mRNA | AK033500.1 | - | 1 | 102 | ||
BC131978.1 | - | 1 | 64 | 40130255 | ||
BC132643.1 | - | 1 | 64 | 40130920 | ||
M17192.1 | - | 1 | 132 |