Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1b21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010642.2 | NP_034772.1 | 3 | 431 | ||
GenBank mRNA | AB039276.1 | - | 3 | 434 | ||
AK144221.1 | - | 3 | 398 | |||
BC012243.1 | - | 3 | 365 | 4167772 | ||
BC145383.1 | - | 3 | 387 | 9053941 |