Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010669.2 | NP_034799.2 | 9 | 1576 | ||
GenBank mRNA | AK009241.1 | - | 6 | 682 | ||
AK137688.1 | - | 9 | 1575 | |||
BC119554.1 | - | 1534 | 40057432 | |||
BC119555.1 | - | 1534 | 40057433 | |||
BC122884.1 | - | 9 | 1533 | |||
BC124556.1 | - | 9 | 1533 | |||
BC139807.1 | - | 1534 | 40057436 |