Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap6-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010672.3 | NP_034802.1 | 1 | 559 | ||
GenBank mRNA | D86420.1 | - | 1 | 557 |