Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291747.1 | NP_001278676.1 | 8 | 955 | ||
NM_011725.4 | NP_035855.1 | 8 | 1003 | |||
GenBank mRNA | AK012549.1 | - | 8 | 994 | ||
AK035341.1 | - | 8 | 972 | |||
AK088930.1 | - | 8 | 907 |