Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVI2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006495.3 | NP_006486.3 | 2 | 292 | ||
XM_005257946.3 | XP_005258003.1 | 3 | 402 | |||
GenBank mRNA | AK297087.1 | - | 3 | 337 | ||
AK312822.1 | - | 2 | 237 | |||
BC005926.1 | - | 2 | 253 | 4043339 | ||
DA677356.1 | - | 2 | 293 |