Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026720.2 | NP_080996.2 | 10, 11 | 1248 | ||
GenBank mRNA | AK089186.1 | - | 10, 11 | 1241 | ||
BC058982.1 | - | 10, 11 | 1159 | 5685059 |