Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028544.1 | NP_082820.1 | 12 | 2844 | ||
GenBank mRNA | AK011557.2 | - | 6 | 927 | ||
AK033276.1 | - | 10 | 2132 | |||
AK156602.1 | - | 12 | 2856 | |||
BC072584.1 | - | 9 | 1798 | |||
BC127059.1 | - | 10 | 1770 | 40109852 | ||
BC150766.1 | - | 12 | 2844 | 9087677 |