Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020769.2 | NP_065820.1 | 4 | 4722 | ||
XM_011530998.2 | XP_011529300.1 | 4 | 4744 | |||
XM_011530999.2 | XP_011529301.1 | 5 | 5593 | |||
XM_017029694.1 | XP_016885183.1 | 3 | 4652 | |||
XM_017029695.1 | XP_016885184.1 | 6 | 5726 | |||
GenBank mRNA | AB037739.1 | - | 4 | 4722 | ||
AY121804.1 | - | 4 | 4722 |