Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rph3al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291159.1 | NP_001278088.1 | 3 | 332 | ||
NM_029548.4 | NP_083824.1 | 2 | 194 | |||
GenBank mRNA | AB112925.1 | - | 2 | 161 | ||
AB158403.1 | - | 2 | 202 | |||
AK018335.1 | - | 3 | 332 | |||
BC127960.1 | - | 2 | 165 | 40134813 | ||
DV651301.1 | - | 1 | 41 | |||
DV660756.1 | - | 3 | 286 |