Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169117.1 | NP_001162588.1 | 2, 3 | 806 | ||
NM_001169118.1 | NP_001162589.1 | 2, 3 | 806 | |||
NM_020860.3 | NP_065911.3 | 2, 3 | 806 | |||
GenBank mRNA | AB040915.1 | - | 2, 3 | 494 | ||
AF328905.1 | - | 2, 3 | 808 | |||
AK096846.1 | - | 2, 3 | 657 | |||
AL136577.1 | - | 2, 3 | 349 | |||
BC136449.1 | - | 2, 3 | 605 | 9020438 | ||
BC146661.1 | - | 2, 3 | 702 | 40148056 | ||
BC152554.1 | - | 2, 3 | 702 | 40148824 | ||
BC171766.1 | - | 2, 3 | 604 | 9054420 |