Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004360.3 | NP_001004360.2 | 2 | 213 | ||
NM_001178034.1 | NP_001171505.1 | 2 | 140 | |||
NM_001178036.1 | NP_001171507.1 | 3 | 617 | |||
NM_001178037.1 | NP_001171508.1 | 2 | 183 | |||
NM_001321905.1 | NP_001308834.1 | 3 | 643 | |||
NM_001321906.1 | NP_001308835.1 | 3 | 259 | |||
NM_001321907.1 | NP_001308836.1 | 2 | 528 | |||
NM_001321909.1 | NP_001308838.1 | 2 | 213 | |||
NM_001321910.1 | NP_001308839.1 | 3 | 586 | |||
NM_001321911.1 | NP_001308840.1 | 2 | 585 | |||
NM_001321912.1 | NP_001308841.1 | 3 | 617 | |||
NM_001321914.1 | NP_001308843.1 | 2 | 143 | |||
NM_020868.4 | NP_065919.2 | 2 | 528 | |||
XM_017004565.1 | XP_016860054.1 | 3 | 687 | |||
GenBank mRNA | AB040925.2 | - | 2 | 95 | ||
AK055256.1 | - | 4 | 701 | |||
AK289967.1 | - | 2 | 227 | |||
AK296490.1 | - | 2 | 215 | |||
AK315147.1 | - | 2 | 332 | |||
AY172661.3 | - | 2 | 107 | |||
AY387785.1 | - | 2 | 129 | |||
BC030832.1 | - | 2 | 564 | 4811771 | ||
BI597037.1 | - | 2 | 170 | |||
DQ857321.1 | - | 2 | 95 | |||
DQ857322.1 | - | 2 | 119 | |||
DQ857323.1 | - | 2 | 86 |