Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314017.1 | NP_001300946.1 | 1, 2 | 170 | ||
NM_020939.1 | NP_065990.1 | 1, 2 | 170 | |||
XM_005249247.2 | XP_005249304.1 | 1, 2 | 170 | |||
XM_011514768.1 | XP_011513070.1 | 1, 2 | 170 | |||
XM_011514769.1 | XP_011513071.1 | 1, 2 | 170 | |||
XM_011514772.1 | XP_011513074.1 | 1, 2 | 170 | |||
GenBank mRNA | AB046819.1 | - | 1, 2 | 728 | ||
DA265730.1 | - | 1, 2 | 170 |