Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174866.2 | NP_777355.1 | 2 | 62 | ||
GenBank mRNA | BC044756.2 | - | 1 | 37 | ||
BC128019.1 | - | 2 | 65 | 40136266 |