Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Szrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025608.2 | NP_001020779.1 | 4 | 1253 | ||
NM_001277195.1 | NP_001264124.1 | 4 | 1250 | |||
GenBank mRNA | AK145912.1 | - | 3 | 1175 | ||
AK171702.1 | - | 3 | 1176 | |||
AK184848.1 | - | 1 | 62 | |||
BC066992.1 | - | 4 | 1201 | 5685985 |