Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020956.2 | NP_066007.1 | 6 | 921 | ||
GenBank mRNA | AF321192.1 | - | 6 | 921 |