Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252094.1 | NP_001239023.1 | 5 | 1563 | ||
NM_001252095.1 | NP_001239024.1 | 5 | 1544 | |||
NM_001252096.1 | NP_001239025.1 | 4 | 1453 | |||
NM_001252097.1 | NP_001239026.1 | 3 | 1025 | |||
NM_177101.5 | NP_796075.2 | 4 | 1434 | |||
GenBank mRNA | AK132096.1 | - | 5 | 1431 | ||
BC018279.1 | - | 2 | 732 | |||
BC046911.1 | - | 3 | 964 | 6491688 | ||
BC048711.1 | - | 4 | 1326 | 6328428 | ||
BC065807.1 | - | 4 | 1350 | 3373106 |