Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl2c5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181852.1 | NP_862900.1 | 4, 5 | 499 | ||
GenBank mRNA | AF128884.1 | - | 4, 5 | 499 | ||
AK145824.1 | - | 5, 6 | 694 | |||
BC132514.1 | - | 4, 5 | 531 | 40130791 | ||
BC132516.1 | - | 4, 5 | 531 | 40130793 |
Gene Symbol: Prl2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045532.2 | NP_001038997.1 | 4, 5 | 559 |