Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029681.3 | NP_083957.1 | 2 | 434 | ||
NM_194065.3 | NP_919041.1 | 2 | 434 | |||
GenBank mRNA | AK007170.1 | - | 2 | 435 | ||
AY369079.1 | - | 2 | 435 | |||
AY369080.1 | - | 2 | 435 | |||
BC114542.1 | - | 2 | 435 | 40050280 |