Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021015.3 | NP_066295.3 | 7 | 611 | ||
NM_175723.1 | NP_783729.1 | 6 | 488 | |||
XM_011543949.2 | XP_011542251.1 | 6 | 523 | |||
XM_017029758.1 | XP_016885247.1 | 6 | 523 | |||
GenBank mRNA | BC016640.2 | - | 6 | 577 | 3923927 | |
BE896093.1 | - | 6 | 591 | |||
U90842.1 | - | 5 | 435 |