Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rexo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207234.2 | NP_997117.2 | 8 | 2109 | ||
GenBank mRNA | AK133982.1 | - | 8 | 2030 | ||
BC060147.1 | - | 8 | 2035 | 6414260 |