Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOBTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242359.1 | NP_001229288.1 | 12 | 2944 | ||
NM_014836.4 | NP_055651.1 | 11 | 2872 | |||
NR_024554.2 | 12 | 2746 | ||||
NR_024555.1 | 12 | 2710 | ||||
XM_006718084.1 | XP_006718147.1 | 11 | 2680 | |||
XM_006718085.1 | XP_006718148.1 | 11 | 2707 | |||
XM_011540424.1 | XP_011538726.1 | 13 | 2620 | |||
XM_011540425.1 | XP_011538727.1 | 11 | 2632 | |||
XM_017017000.1 | XP_016872489.1 | 11 | 2927 | |||
XM_017017001.1 | XP_016872490.1 | 12 | 2805 | |||
XM_017017002.1 | XP_016872491.1 | 10 | 2817 | |||
XM_017017003.1 | XP_016872492.1 | 13 | 2813 | |||
XM_017017004.1 | XP_016872493.1 | 13 | 2815 | |||
XM_017017005.1 | XP_016872494.1 | 12 | 2720 | |||
XM_017017006.1 | XP_016872495.1 | 12 | 2769 | |||
XM_017017007.1 | XP_016872496.1 | 10 | 2819 | |||
XM_017017008.1 | XP_016872497.1 | 13 | 2862 | |||
GenBank mRNA | AB018283.3 | - | 11 | 2763 | ||
AK309440.1 | - | 3 | 808 | |||
BC041791.1 | - | 11 | 2663 | 5297756 |