Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000207.2 | NP_000198.1 | 3 | 441 | ||
NM_001185097.1 | NP_001172026.1 | 3 | 467 | |||
NM_001185098.1 | NP_001172027.1 | 2 | 620 | |||
NM_001291897.1 | NP_001278826.1 | 3 | 501 | |||
GenBank mRNA | BC005255.1 | - | 3 | 441 | 3950204 | |
BM510347.1 | - | 3 | 462 | |||
BM510748.1 | - | 3 | 445 | |||
JQ951950.1 | - | 3 | 426 | |||
X70508.1 | - | 3 | 426 |