Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034173.3 | NP_001029345.2 | 6 | 843 | ||
NR_027752.1 | 6 | 839 | ||||
XM_011537986.2 | XP_011536288.1 | 4 | 402 | |||
XM_011537988.2 | XP_011536290.1 | 6 | 864 | |||
XM_011537989.2 | XP_011536291.1 | 6 | 864 | |||
XM_017018889.1 | XP_016874378.1 | 7 | 1049 | |||
XM_017018890.1 | XP_016874379.1 | 6 | 864 | |||
GenBank mRNA | AK095827.1 | - | 6 | 738 | ||
AK300373.1 | - | 6 | 738 | |||
BC103934.1 | - | 6 | 738 | 40008174 | ||
BC103935.1 | - | 6 | 734 | 40008176 | ||
CR627287.1 | - | 2 | 58 |