Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020992.3 | NP_066272.1 | 2 | 455 | ||
GenBank mRNA | AJ310549.1 | - | 2 | 222 | ||
AK314792.1 | - | 2 | 312 | |||
BC000915.2 | - | 2 | 331 | 2985229 | ||
BC018755.1 | - | 2 | 246 | 3610938 | ||
FJ224331.1 | - | 2 | 246 | |||
U90878.1 | - | 2 | 364 |