Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLT3LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204502.1 | NP_001191431.1 | 5, 6 | 444 | ||
NM_001204503.1 | NP_001191432.1 | 4, 5 | 413 | |||
NM_001278637.1 | NP_001265566.1 | 5, 6 | 472 | |||
NM_001278638.1 | NP_001265567.1 | 5, 6 | 461 | |||
NM_001459.3 | NP_001450.2 | 5, 6 | 444 | |||
XM_011526675.2 | XP_011524977.1 | 5, 6 | 480 | |||
XM_011526676.2 | XP_011524978.1 | 5, 6 | 445 | |||
XM_011526677.2 | XP_011524979.1 | 4, 5 | 434 | |||
XM_011526682.2 | XP_011524984.1 | 4, 5 | 451 | |||
XM_017026533.1 | XP_016882022.1 | 4, 5 | 370 | |||
XM_017026534.1 | XP_016882023.1 | 4, 5 | 343 | |||
GenBank mRNA | AK301136.1 | - | 4, 5 | 405 | ||
BC006331.2 | - | 5, 6 | 434 | |||
BC011914.2 | - | 5, 6 | 434 | |||
BC028001.1 | - | 5, 6 | 445 | 5198812 | ||
BC126293.1 | - | 5, 6 | 401 | 8992009 | ||
BC136464.1 | - | 5, 6 | 401 | 9020453 | ||
BC144129.1 | - | 5, 6 | 401 | 9052649 | ||
U03858.1 | - | 4, 5 | 417 | |||
U04806.1 | - | 5, 6 | 426 |