Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242758.1 | NP_001229687.1 | 8 | 1532 | ||
NM_002116.7 | NP_002107.3 | 8 | 1532 | |||
GenBank mRNA | AB209117.1 | - | 7 | 1615 | ||
AK304472.1 | - | 8 | 1426 | |||
BC019236.2 | - | 8 | 1453 | 2821717 | ||
BE875389.1 | - | 3 | 445 | |||
D32129.1 | - | 8 | 1453 | |||
M24095.1 | - | 7 | 1370 | |||
X02457.1 | - | 7 | 1334 |