Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002584.2 | NP_002575.1 | 8 | 2127 | ||
NM_013945.2 | NP_039236.1 | 8 | 2121 | |||
GenBank mRNA | BC121165.2 | - | 8 | 1603 | 40121582 | |
BC121166.2 | - | 8 | 1603 | 40121584 | ||
X96743.1 | - | 8 | 2128 |