Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002915.3 | NP_002906.1 | 2, 3 | 336 | ||
NM_181558.2 | NP_853536.2 | 2, 3 | 336 | |||
XM_011535172.2 | XP_011533474.1 | 2, 3 | 356 | |||
XM_011535173.2 | XP_011533475.1 | 2, 3 | 356 | |||
XM_011535174.2 | XP_011533476.1 | 2, 3 | 356 | |||
XM_011535175.1 | XP_011533477.1 | 2, 3 | 356 | |||
XM_017020680.1 | XP_016876169.1 | 2, 3 | 356 | |||
XM_017020681.1 | XP_016876170.1 | 2, 3 | 356 | |||
XM_017020682.1 | XP_016876171.1 | 2, 3 | 356 | |||
XM_017020683.1 | XP_016876172.1 | 2, 3 | 356 | |||
GenBank mRNA | AK296527.1 | - | 2, 3 | 277 | ||
AK312544.1 | - | 2, 3 | 252 | |||
AL596753.1 | - | 2, 3 | 364 | |||
BC000149.2 | - | 2, 3 | 267 | 2900958 | ||
BT019668.1 | - | 2, 3 | 226 | |||
CX786577.1 | - | 1, 2 | 78 | |||
DA943219.1 | - | 2, 3 | 336 | |||
L07541.1 | - | 2, 3 | 235 |