Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004197.1 | NP_004188.1 | 7, 8 | 1174 | ||
NM_032454.1 | NP_115830.1 | 7, 8 | 1186 | |||
NR_026717.1 | 7, 8 | 1359 | ||||
GenBank mRNA | AK307198.1 | - | 5, 6 | 610 | ||
BC110347.1 | - | 7, 8 | 1359 | 5165123 | ||
BC121128.1 | - | 6, 7 | 1193 | 40120738 | ||
BC121129.1 | - | 6, 7 | 1192 | 40120740 | ||
BC143833.1 | - | 6, 7 | 1229 | 9052348 | ||
BC143835.1 | - | 6, 7 | 1241 | |||
L26260.1 | - | 7, 8 | 1174 | |||
X77386.1 | - | 6, 7 | 780 |