Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESRRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282450.1 | NP_001269379.1 | 7 | 2235 | ||
NM_001282451.1 | NP_001269380.1 | 7 | 2219 | |||
NM_004451.4 | NP_004442.3 | 7 | 2222 | |||
XM_017017313.1 | XP_016872802.1 | 6 | 2726 | |||
GenBank mRNA | BC007915.2 | - | 6 | 1692 | ||
BC011528.1 | - | 1 | 898 | |||
BC033701.1 | - | 5 | 1631 | |||
BC063795.1 | - | 7 | 2172 | 6527166 | ||
BC073956.1 | - | 3 | 1088 | |||
BC092470.1 | - | 7 | 2235 | 30336569 | ||
L38487.1 | - | 7 | 2175 | |||
X51416.1 | - | 2359 |