Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005645.3 | NP_005636.1 | 1, 2 | 89 | ||
GenBank mRNA | AK312158.1 | - | 1, 2 | 85 | ||
AU280379.1 | - | 1, 2 | 89 | |||
AV713996.1 | - | 1, 2 | 36 | |||
BC121180.1 | - | 1, 2 | 63 | 40123243 | ||
BC121181.1 | - | 1, 2 | 63 | 40123247 | ||
X84003.1 | - | 1, 2 | 45 |