Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318875.1 | NP_001305804.1 | 12 | 1774 | ||
NM_005803.3 | NP_005794.1 | 13 | 1918 | |||
GenBank mRNA | AF089750.2 | - | 13 | 1743 | ||
AF117234.1 | - | 11 | 1376 |