Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006711244.3 | XP_006711307.1 | 7 | 1342 |
Gene Symbol: CCT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282909.1 | NP_001269838.1 | 1, 2 | 263 | ||
NM_006585.3 | NP_006576.2 | 1, 2 | 263 | |||
GenBank mRNA | AK222490.1 | - | 1, 2 | 81 | ||
AK298801.1 | - | 1, 2 | 117 | |||
AK308063.1 | - | 1, 2 | 114 | |||
BC035472.1 | - | 1, 2 | 79 | |||
BC072001.1 | - | 1, 2 | 121 | 6298609 | ||
BC095470.1 | - | 1, 2 | 103 | 30523886 | ||
BP248736.1 | - | 1, 2 | 263 | |||
BU932439.1 | - | 1, 2 | 197 | |||
D13627.1 | - | 1, 2 | 84 |