Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTN3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145008.1 | NP_001138480.1 | 5 | 1039 | ||
NM_001145009.1 | NP_001138481.1 | 5 | 1195 | |||
NM_007048.5 | NP_008979.3 | 5 | 1195 | |||
NM_194441.2 | NP_919423.1 | 5 | 1025 | |||
XM_005248833.2 | XP_005248890.1 | 5 | 1163 | |||
XM_005248834.3 | XP_005248891.1 | 5 | 1190 | |||
XM_006714983.2 | XP_006715046.1 | 4 | 900 | |||
GenBank mRNA | AK223474.1 | - | 4 | 976 | ||
AK290193.1 | - | 5 | 1192 | |||
AK291162.1 | - | 5 | 1025 | |||
AK295720.1 | - | 5 | 1182 | |||
AK298652.1 | - | 5 | 924 | |||
AK299327.1 | - | 5 | 981 | |||
BC118586.1 | - | 5 | 1156 | 40085016 | ||
BC121800.1 | - | 5 | 1156 | 40085015 | ||
BG744194.1 | - | 2 | 258 | |||
BM542639.1 | - | 568 | ||||
U90552.2 | - | 5 | 1125 | |||
Y07827.1 | - | 5 | 917 |