Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314030.1 | NP_001300959.1 | 3 | 657 | ||
NM_012284.2 | NP_036416.1 | 3 | 661 | |||
XM_011538085.2 | XP_011536387.1 | 3 | 1277 | |||
XM_011538086.2 | XP_011536388.1 | 3 | 1273 | |||
XM_017019096.1 | XP_016874585.1 | 2 | 1043 | |||
XM_017019097.1 | XP_016874586.1 | 2 | 206 | |||
GenBank mRNA | AB022696.1 | - | 3 | 436 | ||
AB033108.1 | - | 3 | 661 | |||
BC150602.1 | - | 3 | 542 | 9056972 |