Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014248.3 | NP_055063.1 | 1, 2 | 129 | ||
GenBank mRNA | AF085906.1 | - | 1, 2 | 58 | ||
AF140598.1 | - | 1, 2 | 83 | |||
AF142059.1 | - | 1, 2 | 77 | |||
AK315722.1 | - | 1, 2 | 88 | |||
AY099360.1 | - | 1, 2 | 40 | |||
BC001466.2 | - | 1, 2 | 82 | 3138751 | ||
BC017370.1 | - | 1, 2 | 99 | |||
CR456560.1 | - | 1, 2 | 95 | |||
DA353856.1 | - | 1, 2 | 129 |