Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF644 SIRNA Assays ›
Gene Symbol:
ZNF644
Gene Name:
zinc finger protein 644
Gene Aliases:
BM-005, MYP21, NatF, ZEP-2
Chromosome Location:
Chr.1: 90915298 - 91022269 on Build GRCh38
UniGene ID:
Hs.173001
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
289773
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF644
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_016620.3
NP_057704.2
3
324
NM_032186.4
NP_115562.3
3
354
NM_201269.2
NP_958357.1
5
3998
XM_005271257.4
XP_005271314.1
7
4457
XM_005271260.4
XP_005271317.1
6
4434
XM_011542258.2
XP_011540560.1
6
4135
XM_011542259.2
XP_011540561.1
8
4811
XM_011542260.2
XP_011540562.1
8
4715
XM_011542261.2
XP_011540563.1
6
4056
XM_017002487.1
XP_016857976.1
7
4246
XM_017002488.1
XP_016857977.1
6
4334
XM_017002489.1
XP_016857978.1
5
4309
XM_017002490.1
XP_016857979.1
6
4121
XM_017002491.1
XP_016857980.1
5
3933
XM_017002492.1
XP_016857981.1
4
1344
XM_017002493.1
XP_016857982.1
4
690
XM_017002494.1
XP_016857983.1
4
467
GenBank mRNA
AB033047.1
-
3
3581
AK001587.1
-
3
1763
AK023596.1
-
3
937
AK291520.1
-
3
124
BC031003.1
-
2
385
BC050656.2
-
3
273
BC063683.1
-
4
3692
BC110841.1
-
3
172
BC132776.1
-
5
3813
40146798
BC132778.1
-
5
3813
40146800
BC150177.1
-
5
3953
40148078
BI760689.1
-
320
DA147194.1
-
3
326
DB001551.1
-
3
463
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0