Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UCKL1 SIRNA Assays ›
Gene Symbol:
UCKL1
Gene Name:
uridine-cytidine kinase 1 like 1
Gene Aliases:
UCK1L, URKL1
Chromosome Location:
Chr.20: 63939829 - 63959435 on Build GRCh38
UniGene ID:
Hs.504998
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
289792
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UCKL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001193379.1
NP_001180308.1
15
2007
NM_017859.3
NP_060329.2
15
1779
NR_126526.1
15
1756
XM_005260216.2
XP_005260273.1
15
1747
XM_006723806.2
XP_006723869.1
15
1753
XM_006723807.2
XP_006723870.1
15
1750
XM_006723808.1
XP_006723871.1
15
1805
XM_006723809.1
XP_006723872.1
15
1785
XM_006723810.1
XP_006723873.1
15
1782
XM_006723811.3
XP_006723874.1
15
3774
XM_011528868.2
XP_011527170.1
15
3777
XM_011528869.1
XP_011527171.1
16
5048
XM_011528870.1
XP_011527172.1
15
1767
XM_011528871.1
XP_011527173.1
15
1794
XM_011528872.1
XP_011527174.1
16
5037
XM_017027896.1
XP_016883385.1
15
1782
XM_017027898.1
XP_016883387.1
15
1779
XM_017027899.1
XP_016883388.1
15
1776
XM_017027900.1
XP_016883389.1
15
2628
XM_017027901.1
XP_016883390.1
15
2623
XM_017027902.1
XP_016883391.1
16
1985
XM_017027903.1
XP_016883392.1
15
1788
XM_017027904.1
XP_016883393.1
15
4709
XM_017027905.1
XP_016883394.1
15
4706
XM_017027906.1
XP_016883395.1
15
4703
XM_017027907.1
XP_016883396.1
11
1505
XM_017027908.1
XP_016883397.1
12
1408
GenBank mRNA
AK222559.1
-
15
1750
AK223220.1
-
15
1748
BC033078.1
-
13
1766
4810505
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide kinase
Gene Ontology Categories:
Function(s)
Process(es)
pyrimidine nucleobase metabolic process
viral process
phosphorylation
pyrimidine nucleoside salvage
UMP salvage
CTP salvage
uridine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0