Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMBR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300750.1 | NP_001287679.1 | 18 | 2512 | ||
NM_001300751.1 | NP_001287680.1 | 17 | 2250 | |||
NM_018113.3 | NP_060583.2 | 17 | 2310 | |||
XM_011538563.2 | XP_011536865.1 | 17 | 2464 | |||
XM_011538564.2 | XP_011536866.1 | 17 | 2837 | |||
XM_011538565.2 | XP_011536867.1 | 16 | 3086 | |||
XM_011538566.2 | XP_011536868.1 | 16 | 2060 | |||
XM_017019626.1 | XP_016875115.1 | 17 | 2712 | |||
XM_017019627.1 | XP_016875116.1 | 18 | 2441 | |||
XM_017019628.1 | XP_016875117.1 | 16 | 2137 | |||
XM_017019629.1 | XP_016875118.1 | 18 | 3296 | |||
XM_017019630.1 | XP_016875119.1 | 14 | 4272 | |||
XM_017019631.1 | XP_016875120.1 | 16 | 2000 | |||
GenBank mRNA | AB033000.1 | - | 15 | 2781 | ||
AF260728.1 | - | 17 | 2269 | |||
AK001356.1 | - | 18 | 2293 | |||
AK001651.1 | - | 17 | 2200 | |||
AK093570.1 | - | 11 | 2043 | |||
AL137599.1 | - | 14 | 1703 | |||
AY358572.1 | - | 17 | 2274 | |||
BC008337.1 | - | 17 | 2180 | 3506347 | ||
BC015015.1 | - | 17 | 2039 | 3910520 | ||
BC031550.1 | - | 17 | 2071 | 5194023 |