Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PBRM1 SIRNA Assays ›
Gene Symbol:
PBRM1
Gene Name:
polybromo 1
Gene Aliases:
BAF180, PB1
Chromosome Location:
Chr.3: 52545352 - 52685933 on Build GRCh38
UniGene ID:
Hs.189920
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
289797
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PBRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_018313.4
NP_060783.3
13
1535
XM_005265279.4
XP_005265336.1
14
1763
XM_005265280.3
XP_005265337.1
13
1580
XM_005265282.3
XP_005265339.1
14
1722
XM_005265283.3
XP_005265340.1
13
1577
XM_011533900.2
XP_011532202.1
16
1929
XM_011533902.2
XP_011532204.1
15
1861
XM_011533903.2
XP_011532205.1
15
1866
XM_017006725.1
XP_016862214.1
15
1836
XM_017006726.1
XP_016862215.1
16
1946
XM_017006727.1
XP_016862216.1
16
1891
XM_017006728.1
XP_016862217.1
15
1887
XM_017006729.1
XP_016862218.1
15
2295
XM_017006730.1
XP_016862219.1
16
1882
XM_017006731.1
XP_016862220.1
15
1865
XM_017006732.1
XP_016862221.1
15
1861
XM_017006733.1
XP_016862222.1
15
2255
XM_017006734.1
XP_016862223.1
15
1828
XM_017006735.1
XP_016862224.1
15
2294
XM_017006736.1
XP_016862225.1
15
2295
XM_017006737.1
XP_016862226.1
14
2169
XM_017006738.1
XP_016862227.1
14
1790
XM_017006739.1
XP_016862228.1
14
2166
XM_017006740.1
XP_016862229.1
14
1707
XM_017006741.1
XP_016862230.1
15
1843
XM_017006742.1
XP_016862231.1
15
1866
XM_017006743.1
XP_016862232.1
15
2295
XM_017006744.1
XP_016862233.1
15
1832
XM_017006745.1
XP_016862234.1
14
2141
XM_017006746.1
XP_016862235.1
14
2138
XM_017006747.1
XP_016862236.1
14
2200
XM_017006748.1
XP_016862237.1
14
2141
XM_017006749.1
XP_016862238.1
14
1707
XM_017006750.1
XP_016862239.1
13
1595
XM_017006751.1
XP_016862240.1
15
1843
XM_017006752.1
XP_016862241.1
15
2295
XM_017006753.1
XP_016862242.1
15
2294
XM_017006754.1
XP_016862243.1
15
1861
XM_017006755.1
XP_016862244.1
15
1843
XM_017006756.1
XP_016862245.1
15
1843
XM_017006757.1
XP_016862246.1
13
1566
XM_017006758.1
XP_016862247.1
13
1697
XM_017006759.1
XP_016862248.1
15
1843
XM_017006760.1
XP_016862249.1
15
1861
XM_017006761.1
XP_016862250.1
15
1870
XM_017006762.1
XP_016862251.1
15
1861
XM_017006763.1
XP_016862252.1
16
1920
XM_017006764.1
XP_016862253.1
15
1843
XM_017006765.1
XP_016862254.1
14
1722
XM_017006766.1
XP_016862255.1
15
1850
GenBank mRNA
AF177387.1
-
12
1424
AF197569.1
-
13
1517
AF225870.1
-
11
1328
AF225871.1
-
12
1424
AF225872.1
-
12
1424
AK056541.1
-
11
1253
AY281068.1
-
12
1423
BC115009.1
-
12
1439
7962003
BC115010.1
-
12
1440
BC115011.1
-
12
1440
7962010
BC129934.1
-
12
1424
40082629
BC129935.1
-
12
1424
40082630
BU171600.1
-
5
630
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
negative regulation of cell proliferation
covalent chromatin modification
DNA binding
chromatin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0