Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018419.2 | NP_060889.1 | 2 | 1581 | ||
GenBank mRNA | AF086136.1 | - | 417 | |||
AF270652.1 | - | 2 | 1593 | |||
AJ243896.1 | - | 2 | 1483 | |||
BC035513.1 | - | 2 | 1581 | |||
BC038590.1 | - | 2 | 1583 | 5589289 | ||
BC073971.1 | - | 2 | 1583 | 6183010 | ||
BC111390.1 | - | 2 | 1581 | 5751646 |