Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTS2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018949.2 | NP_061822.1 | 1 | 313 | ||
GenBank mRNA | BC136490.1 | - | 1 | 313 | 9020479 |
Set Membership: |
G-protein coupled receptor |